missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Citric Acid U.S.P. Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 5949-29-1
Physical Form Granular
Molecular Formula C6H8O7·H2O
Formula Weight 210.14
Chemical Name or Material Citric Acid, Monohydrate
Grade USP
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK0616212 Avantor Performance
061606
Polyethylene 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

5949-29-1
C6H8O7·H2O
Citric Acid, Monohydrate
Granular
210.14
USP

Specifications

5949-29-1
C6H8O7·H2O
Polyethylene
Citric Acid, Monohydrate
Granular
210.14
2.5kg
USP