Please login to your online account to display your discounted pricing

Citric Acid, U.S.P. - F.C.C., J.T.Baker™

$679.43 - $679.43

Chemical Identifiers

Chemical Name or Material Citric Acid
CAS 5949-29-1
Purity Grade FCC/USP
Molecular Formula HOC(COOH)(CH2COOH)2·H2O
Formula Weight 210.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002609

 
avantor performance
011905
Poly Bottle 2.5kg Each for $679.43
Description & Specifications

Specifications

Purity Grade FCC/USP
Formula Weight 210.14
Chemical Name or Material Citric Acid
CAS 5949-29-1
Molecular Formula HOC(COOH)(CH2COOH)2·H2O
Packaging Poly Bottle
Quantity 2.5kg
Specific Gravity 1L = 1.54kg