missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Citric acid, trisodium salt dihydrate, ACS reagent, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 446330050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Citric acid, trisodium salt dihydrate | |
Plastic bottle | |
NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O | |
15,8737 | |
Solubility in water: 720g/L (25°C). Other solubilities: insoluble in alcohol | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+] | |
294.09 | |
CHEBI:32142 | |
ACS Reagent |
6132-04-3 | |
C6H5Na3O7·2H2O | |
5kg | |
trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate | |
NLJMYIDDQXHKNR-UHFFFAOYSA-K | |
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate | |
71474 | |
294.09 | |
0.005% max |
RUO â Research Use Only