missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Citric acid, trisodium salt dihydrate, 99%, pure, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC227130250
Specifications
Citric acid, trisodium salt dihydrate | |
99% | |
NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O | |
15, 8737 | |
Solubility in water: 720g/L (25°C). Other solubilities: insoluble in alcohol | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+] | |
294.10 | |
CHEBI:32142 | |
Crystals or Crystalline Powder | |
Pure | |
Fibre drum | |
11 to 13% | |
White |
6132-04-3 | |
C6H9Na3O9 | |
MFCD00150031 | |
1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, citronensaeure-tri-na-salz-dihydrat, natriumcitrat-dihydrat, sodium citrate dihydrate, sodium citrate hydrate, sodium citrate tribasic dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, trisodium citrate dihydrate | |
NLJMYIDDQXHKNR-UHFFFAOYSA-K | |
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate | |
71474 | |
294.09 | |
99% | |
Authentic | |
Solubility in water: 720g/L (25°C). Other solubilities: insoluble in alcohol | |
>300.0°C | |
25kg |
Chemical Identifiers
6132-04-3 | |
294.10 | |
NLJMYIDDQXHKNR-UHFFFAOYSA-K | |
71474 | |
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate |
C6H9Na3O9 | |
MFCD00150031 | |
1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, citronensaeure-tri-na-salz-dihydrat, natriumcitrat-dihydrat, sodium citrate dihydrate, sodium citrate hydrate, sodium citrate tribasic dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, trisodium citrate dihydrate | |
CHEBI:32142 | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+] |
Safety and Handling
Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated.
NFPA
Health:
Signal Word: CAUTION!