missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Citric acid, trisodium salt 98%, anhydrous, pure, ACROS Organics™

Manufacturer:  Fisher Scientific Acros AC391971000

 View more versions of this product

Catalog No. AC391971000


 

Specifications Description & Specifications

Specifications

Citric acid, trisodium salt
Pure
≥97.5%
C6H5Na3O7
MFCD00012462
Trisodium citrate
Plastic bottle
100g
68-04-2
Crystalline Powder
98%
258.07
15, 8737
Authentic
White to off-white

Chemical Identifiers

68-04-2
C6H5Na3O7
MFCD00012462
Citric acid, trisodium salt
Pure
98%
258.07
Trisodium citrate
≥97.5%