missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Citric acid, trisodium salt 98%, anhydrous, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 391970025

 View more versions of this product

Catalog No. AC391970025



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Citric acid, trisodium salt
300.0°C
290°C
0.98
C6H5Na3O7
2.5 kg
sodium citrate, trisodium citrate, citrosodine, sodium citrate anhydrous, citrosodina, natrocitral, citnatin, citreme, citrosodna, trisodium citrate, anhydrous
HRXKRNGNAMMEHJ-UHFFFAOYSA-K
trisodium;2-hydroxypropane-1,2,3-tricarboxylate
6224
258.07
Pure
68-04-2
White
Authentic
Plastic bottle
MFCD00012462
15, 8737
Solubility in water: 500g/L (20°C).
[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
258.07
CHEBI:53258
0.98
Crystalline Powder

Chemical Identifiers

68-04-2
258.07
HRXKRNGNAMMEHJ-UHFFFAOYSA-K
6224
trisodium;2-hydroxypropane-1,2,3-tricarboxylate
C6H5Na3O7
MFCD00012462
sodium citrate, trisodium citrate, citrosodine, sodium citrate anhydrous, citrosodina, natrocitral, citnatin, citreme, citrosodna, trisodium citrate, anhydrous
CHEBI:53258
[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only