Please login to your online account to display your discounted pricing

Citric acid, triammonium salt, +97%, pure, ACROS Organics™


Quantity 50g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC391410500

 View more versions of this product

Catalog No. AC391410500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.0%
Purity Grade Extra Pure
Physical Form Crystals
Formula Weight 243.22
Quantity 50g
Color White to off-white
Chemical Name or Material Citric acid, triammonium salt
Assay Percent Range 97+%
MDL Number MFCD00036406
CAS 3458-72-8
Infrared Spectrum Authentic
Packaging Plastic bottle
Molecular Formula C6H17N3O7
Synonym Triammoniumcitrate
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Citric acid, triammonium salt
CAS 3458-72-8
Purity Grade Extra Pure
Assay ≥97.0%
Assay Percent Range 97+%
Molecular Formula C6H17N3O7
Formula Weight 243.22
MDL Number MFCD00036406
Synonym Triammoniumcitrate
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents