Please login to your online account to display your discounted pricing

Citric acid, monosodium salt, 99%, pure, anhydrous, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC215330025

 View more versions of this product

Catalog No. AC215330025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Purity Grade Extra Pure
Physical Form Crystalline Powder
Formula Weight 214.11
Quantity 2.5kg
Color White
TSCA TSCA
EINECS Number 242-734-6
RTECS Number GE7950000
Beilstein 03, 563
Infrared Spectrum Authentic
Chemical Name or Material Citric acid, monosodium salt
Molecular Formula C6H7NaO7
Linear Formula HO2CCH2C(OH)(CO2H)CH2CO2Na
MDL Number MFCD00013067
CAS 18996-35-5
Name Note 99%
Assay Percent Range 99%
Packaging Plastic bottle
Synonym Sodium dihydrogen citrate
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 HYGROSCOPIC

Specifications

Chemical Name or Material Citric acid, monosodium salt
Name Note 99%
CAS 18996-35-5
Purity Grade Extra Pure
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H7NaO7
Linear Formula HO2CCH2C(OH)(CO2H)CH2CO2Na
Formula Weight 214.11
MDL Number MFCD00013067
Synonym Sodium dihydrogen citrate
Accessories

HYGROSCOPIC

GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.

SDS & Safety Documents