missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Citric Acid, For Radium, Certified, 1.00M ±0.01M (3.0N), LabChem™

Manufacturer:  LabChem LC131802

 View more versions of this product

Catalog No. LC131802



Specifications

Specifications

Citric Acid
77-92-9,7732-18-5
C6H8O7
1.00M ±0.01M (3.0N)
KRKNYBCHXYNGOX-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid
311
192.12
Certified
Poly Bottle
Room Temperature
Fume; Carbon monoxide; Carbon dioxide
1L
For Radium
80.8,19.2
C6H8O7
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
C(C(=O)O)C(CC(=O)O)(C(=O)O)O
192.123
CHEBI:30769
Liquid
Passes Test
Soluble in water
Traceable to NIST
Colorless

Chemical Identifiers

77-92-9
192.123
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
CHEBI:30769
C(C(=O)O)C(CC(=O)O)(C(=O)O)O
C6H8O7
KRKNYBCHXYNGOX-UHFFFAOYSA-N
311
2-hydroxypropane-1,2,3-tricarboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Documents
Promotions

Promotions