Learn More
Citric acid, diammonium salt, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 392505000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Citric acid, diammonium salt | |
White | |
Authentic | |
Plastic bottle | |
MFCD00013068 | |
1.5 | |
ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss | |
YXVFQADLFFNVDS-UHFFFAOYSA-N | |
azane;2-hydroxypropane-1,2,3-tricarboxylic acid | |
13710713 | |
226.19 | |
Pure |
3012-65-5 | |
1.5000g/mL | |
0.98 | |
C6H14N2O7 | |
500 g | |
15, 509 | |
Solubility in water: soluble. | |
[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O | |
226.19 | |
CHEBI:63076 | |
0.98 | |
Crystalline Powder |
Chemical Identifiers
3012-65-5 | |
226.19 | |
YXVFQADLFFNVDS-UHFFFAOYSA-N | |
13710713 | |
azane;2-hydroxypropane-1,2,3-tricarboxylic acid |
C6H14N2O7 | |
MFCD00013068 | |
ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss | |
CHEBI:63076 | |
[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only