Please login to your online account to display your discounted pricing

Citric Acid, Anhydrous, ACS Grade, ≥ 99.5%, LabChem™

$285.29 - $285.29

Chemical Identifiers

Chemical Name or Material Citric Acid
CAS 77-92-9
CAS Max % 100
Name Note Anhydrous
Purity Grade ACS Grade
Linear Formula C6H8O7
Formula Weight 192.12
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC131401

 
lab chem
LC131401
500g Each for $285.29
Description & Specifications

Specifications

Assay ≥99.5%
Formula Weight 192.12
Chemical Name or Material Citric Acid
Specific Gravity 1.665g/cm3
Solubility Soluble in water
EINECS Number 201-069-1
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Melting Point 153°C
Color White
Boiling Point 175°C
Quantity 500g
Name Note Anhydrous
Purity Grade ACS Grade
Physical Form Crystals
Carbonizable Substances Passes Test
Linear Formula C6H8O7
CAS 77-92-9
Density 1.665g/cm3
Health Hazard 1 Warning
CAS Max % 100
Health Hazard 2 GHS H Statement
Causes serious eye irritation.
Identification Passes Test
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Packaging Poly Bottle
Recommended Storage Room Temperature
Safety and Handling

Warning

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes serious eye irritation.

Room Temperature