Please login to your online account to display your discounted pricing

Citric Acid, 10% (w/v) Aqueous Solution, Ricca Chemical


Quantity 4L
Packaging Poly natural

Manufacturer: ricca chemical company  RI21201

 View more versions of this product

Catalog No. RC21201

  • / Each
 

Description & Specifications

Chemical Identifiers

Color Clear colorless
Physical Form Liquid
Quantity 4L
Solvent or Matrix Aqueous Solution
Concentration or Composition Notes 10% (w/v)
Chemical Name or Material Citric Acid
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 93.0
CAS 77-92-9
CAS Min % 9.0
Packaging Poly natural
CAS Max % 11.0

Specifications

Chemical Name or Material Citric Acid
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 93.0
CAS 77-92-9
CAS Min % 9.0
CAS Max % 11.0
Physical Form Liquid