Please login to your online account to display your discounted pricing

Alfa Aesar™ Citrazinic acid, 97%

$35.27 - $158.22

Chemical Identifiers

Chemical Name or Material Citrazinic acid
CAS 99-11-6
Molecular Formula C6H5NO4
Formula Weight 155.11
MDL Number MFCD00006274
Beilstein 383736
Merck Index 14,2325
Synonym 2,6-Dihydroxyisonicotinic acid; 2,6-Dihydroxypyridine-4-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1546118

  SDS
alfa aesar™
A1546118
50g Each for $35.27

AAA1546130

  SDS
alfa aesar™
A1546130
250g Each for $158.22
Description & Specifications

Specifications

Formula Weight 155.11
Chemical Name or Material Citrazinic acid
Assay 97%
CAS 99-11-6
Merck Index 14,2325
Melting Point >300°C (dec.)
Health Hazard 1 Warning
Quantity 50g
Odor Odorless
MDL Number MFCD00006274
Synonym 2,6-Dihydroxyisonicotinic acid; 2,6-Dihydroxypyridine-4-carboxylic acid
Beilstein 383736
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS Number NS1400000
TSCA TSCA
Molecular Formula C6H5NO4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
2,6-Dihydroxypyridine-4-carboxylic acid was used to prepare 2,6-dibromo-4-(hexoxymethyl)pyridine. It was also used to synthesize the derivatives of oxazinones and pyrimidinones.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.