missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Citrazinic acid, 97%

Manufacturer:  Alfa Aesar™ A1546130

 View more versions of this product

Catalog No. AAA1546130



Description

Description

Applications
2,6-Dihydroxypyridine-4-carboxylic acid was used to prepare 2,6-dibromo-4-(hexoxymethyl)pyridine. It was also used to synthesize the derivatives of oxazinones and pyrimidinones.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

Citrazinic acid
>300°C (decomposition)
99-11-6
MFCD00006274
14,2325
CSGQJHQYWJLPKY-UHFFFAOYSA-N
2-hydroxy-6-oxo-1H-pyridine-4-carboxylic acid
7425
97%
Odorless
250g
C6H5NO4
383736
citrazinic acid, 2,6-dihydroxyisonicotinic acid, 2,6-dihydroxypyridine-4-carboxylic acid, 2,6-dihydroxy-4-carboxypyridine, 2,6-dihydroxy-4-pyridinecarboxylic acid, kyselina citrazinova, 2,6-dihydroxyisonicotinate, 4-pyridinecarboxylic acid, 2,6-dihydroxy, citrazinicacid, 4-pyridinecarboxylic acid, 1,2-dihydro-6-hydroxy-2-oxo
C1=C(C=C(NC1=O)O)C(=O)O
155.109
155.11

Chemical Identifiers

99-11-6
155.109
CSGQJHQYWJLPKY-UHFFFAOYSA-N
7425
C1=C(C=C(NC1=O)O)C(=O)O
C6H5NO4
MFCD00006274
citrazinic acid, 2,6-dihydroxyisonicotinic acid, 2,6-dihydroxypyridine-4-carboxylic acid, 2,6-dihydroxy-4-carboxypyridine, 2,6-dihydroxy-4-pyridinecarboxylic acid, kyselina citrazinova, 2,6-dihydroxyisonicotinate, 4-pyridinecarboxylic acid, 2,6-dihydroxy, citrazinicacid, 4-pyridinecarboxylic acid, 1,2-dihydro-6-hydroxy-2-oxo
2-hydroxy-6-oxo-1H-pyridine-4-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : NS1400000

TSCA : TSCA

Documents