Please login to your online account to display your discounted pricing

Citrate-Cyanide Reducing Solution, For Lead, LabChem™

$69.57 - $69.57

Chemical Identifiers

Chemical Name or Material Citrate-Cyanide Reducing Solution
CAS 1336-21-6
CAS Max % 66.7
CAS 7732-18-5
CAS Max % 17.64
CAS 3012-65-5
CAS Max % 13.33
CAS 151-50-8
CAS Max % 1.33
CAS 7757-83-7
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC131301

 
lab chem
LC131301
500mL Each for $69.57
Description & Specifications

Specifications

Formula Weight 65.12
Chemical Name or Material Citrate-Cyanide Reducing Solution
CAS Max % 13.33
Decomposition Information Hydrogen cyanide; Carbon monoxide; Carbon dioxide; Gaseous Ammonia
CAS 151-50-8
Color Clear, colorless to slight yellow
Quantity 500mL
CAS Max % 1.33
CAS 7757-83-7
CAS Max % 0.67
Name Note For Lead
Physical Form Liquid
Linear Formula KCN
CAS 1336-21-6
UN Number UN1935
Health Hazard 1 Danger
CAS Max % 66.7
Health Hazard 2 GHS H Statement
Toxic if swallowed.
Harmful in contact with skin or if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Suspected of causing cancer.
Identification Passes Test
CAS 7732-18-5
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Contaminated work clothing should not be allowed out of the workplace.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
CAS Max % 17.64
Solubility Soluble in water
CAS 3012-65-5
Storage Information Room Temperature
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Contaminated work clothing should not be allowed out of the workplace.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Toxic if swallowed.
Harmful in contact with skin or if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Suspected of causing cancer.