Please login to your online account to display your discounted pricing

Alfa Aesar™ Citrate Buffer, 0.2M buffer soln., pH 6.0, low endotoxin

Chemical Identifiers

Chemical Name or Material Citrate Buffer
CAS 6132-04-3
Name Note low endotoxin
MDL Number MFCD00150031
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67266AE

 
alfa aesar™
J67266AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67266AK

 
alfa aesar™
J67266AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Citrate Buffer
CAS 6132-04-3
Physical Form Liquid
TSCA TSCA
Quantity 100mL
Storage Note 1 2° to 8°C
pH 6.0
Concentration 0.2M buffer soln.
Color Colorless
Name Note low endotoxin
MDL Number MFCD00150031
Safety and Handling

2° to 8°C