Please login to your online account to display your discounted pricing

Alfa Aesar™ Citrate Buffer, 0.2M buffer soln., pH 5.0, low endotoxin

Chemical Identifiers

Chemical Name or Material Citrate Buffer
CAS 6132-04-3
Name Note low endotoxin
MDL Number MFCD00150031
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67180AE

 
alfa aesar™
J67180AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67180AK

 
alfa aesar™
J67180AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Citrate Buffer
Physical Form Liquid
MDL Number MFCD00150031
CAS 6132-04-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
pH 5.0
Color Colorless
Quantity 100mL
TSCA TSCA
Name Note low endotoxin
Odor Odorless
Storage Note 1 2° to 8°C
Concentration 0.2M buffer soln.
Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.

2° to 8°C