missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Citrate Buffer, 0.2M buffer soln., pH 4.0, low endotoxin
CAS: 6132-04-3 | C6H9Na3O9 | 294.10 g/mol
Supplier: Thermo Scientific Chemicals J67435AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Citrate Buffer | |
6132-04-3 | |
0.2 M | |
Liquid | |
C6H9Na3O9 | |
NLJMYIDDQXHKNR-UHFFFAOYSA-K | |
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate | |
71474 |
low endotoxin | |
Colorless | |
4.0 | |
100 mL | |
MFCD00150031 | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+] | |
294.10 | |
CHEBI:32142 |
RUO – Research Use Only