Please login to your online account to display your discounted pricing

Alfa Aesar™ Citraconic hydrazide, 97%

$46.49 - $571.07

Chemical Identifiers

Chemical Name or Material Citraconic hydrazide
CAS 5754-18-7
Molecular Formula C5H6N2O2
Formula Weight 126.11
MDL Number MFCD00067746
Synonym 1,2-Dihydro-4-methyl-3,6-pyridazinedione; alpha-Methylmaleic hydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2515203

 
alfa aesar™
B2515203
1g Each for $46.49

AAB2515214

 
alfa aesar™
B2515214
25g Each for $571.07
Description & Specifications

Specifications

Formula Weight 126.11
Chemical Name or Material Citraconic hydrazide
Assay 97%
Molecular Formula C5H6N2O2
MDL Number MFCD00067746
CAS 5754-18-7
Synonym 1,2-Dihydro-4-methyl-3,6-pyridazinedione; alpha-Methylmaleic hydrazide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 284° to 288°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.