missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Citraconic Anhydride, TCI America™

Manufacturer:  TCI America M0365500G

 View more versions of this product

Catalog No. M0365500G



Specifications

Specifications

Citraconic Anhydride
Yellow
616-02-4
MFCD00005522
citraconic anhydride, 2,5-furandione, 3-methyl, 2-methylmaleic anhydride, methylmaleic anhydride, 3-methylmaleic anhydride, citraconic acid anhydride, monomethylmaleic anhydride, maleic anhydride, methyl, 2-methylmaleicanhydride, unii-105np7pmxx
CC1=CC(=O)OC1=O
112.084
112.08
≥98.0% (GC)
215°C
500g
C5H4O3
2810
AYKYXWQEBUNJCN-UHFFFAOYSA-N
3-methylfuran-2,5-dione
12012
Clear Liquid at 20°C

Chemical Identifiers

616-02-4
112.084
AYKYXWQEBUNJCN-UHFFFAOYSA-N
12012
CC1=CC(=O)OC1=O
C5H4O3
MFCD00005522
citraconic anhydride, 2,5-furandione, 3-methyl, 2-methylmaleic anhydride, methylmaleic anhydride, 3-methylmaleic anhydride, citraconic acid anhydride, monomethylmaleic anhydride, maleic anhydride, methyl, 2-methylmaleicanhydride, unii-105np7pmxx
3-methylfuran-2,5-dione
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6177

RTECSNumber : GE6825000

TSCA : Yes

Documents