Learn More
Citraconic anhydride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 154790010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Citraconic anhydride | |
616-02-4 | |
213°C | |
Authentic | |
Glass bottle | |
1.4700 to 1.472 | |
1kg | |
1.245 | |
Solubility in water: decomposes in water. Other solubilities: soluble in ethanol and ether | |
CC1=CC(=O)OC1=O | |
112.08 | |
112.08 |
98% | |
1.2450g/mL | |
101°C | |
98% | |
C5H4O3 | |
MFCD00005522 | |
17, 440 | |
citraconic anhydride, 2,5-furandione, 3-methyl, 2-methylmaleic anhydride, methylmaleic anhydride, 3-methylmaleic anhydride, citraconic acid anhydride, monomethylmaleic anhydride, maleic anhydride, methyl, 2-methylmaleicanhydride, unii-105np7pmxx | |
AYKYXWQEBUNJCN-UHFFFAOYSA-N | |
3-methylfuran-2,5-dione | |
12012 | |
98% |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publicat
GHS Signal Word: Danger
EINECSNumber : 210-459-0
RTECSNumber : GE6825000
TSCA : TSCA
RUO â Research Use Only