Please login to your online account to display your discounted pricing

Tocris™ CITCO

Selective CAR agonist

$168.15 - $697.40

Specifications

Product Type CITCO
Inhibitors CITCO
Molecular Weight 436.74
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

368310R

 
tocris™
368310
10mg Each for $168.15

368350R

 
tocris™
368350
50mg Each for $697.40
Description & Specifications

Specifications

Product Type CITCO
Inhibitors CITCO
Molecular Weight 436.74
Purity >98

Constitutive androstane receptor (CAR) agonist (EC50 = 49nM). Displays > 100-fold selectivity over PXR receptors and no activity at LXR, ERα, ERβ, PPAR, RAR, FXR, VDR and THR. Induces CAR nuclear translocation and expression of CYP2B6 in hepatocytes in vitro.

  • Chemical Name: 6-(4-Chlorophenyl)imidazo[2,1-b][1,3]thiazole-5-carbaldehyde-O-(3,4-dichlorobenzyl)oxime
  • Formula: C19H12Cl3N3OS
  • CAS Number: 338404-52-7
  • Solubility: Soluble to 50mM in DMSO and to 5mM in ethanol
  • Purity: >98