Please login to your online account to display your discounted pricing

Cisapride, Tocris

5-HT4 agonist; stimulates intestinal ACh release

$151.85 - $644.39

Chemical Identifiers

Chemical Name or Material Cisapride
Assay Percent Range >99%
Molecular Formula C23H29ClFN3O4
Formula Weight 465.95
Synonym (±)-4-Amino-5-chloro-N-[1-[(3R™, 4S™)-3-(4-fluorophenoxy)propyl]-3-methoxy-4-piperidinyl]-2-methoxybenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

169550

 
r & d systems
RDS169550
50mg Each for $644.39

169510

 
r & d systems
RDS169510
10mg Each for $151.85
Description & Specifications

Specifications

Formula Weight 465.95
Quantity 50mg
Chemical Name or Material Cisapride
CAS 81098-60-4
Assay Percent Range >99%
Molecular Formula C23H29ClFN3O4
Synonym (±)-4-Amino-5-chloro-N-[1-[(3R™, 4S™)-3-(4-fluorophenoxy)propyl]-3-methoxy-4-piperidinyl]-2-methoxybenzamide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature