Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-Stilbeneboronic acid pinacol ester, 99%

$76.50 - $210.88

Chemical Identifiers

Chemical Name or Material cis-Stilbeneboronic acid pinacol ester
CAS 264144-59-4
Molecular Formula C20H23BO2
Formula Weight 306.22
MDL Number MFCD03788734
Beilstein 9269561
Synonym cis-(1,2-Diphenylethenyl)boronic acid pinacol cyclic ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL19576MD

  SDS
alfa aesar™
L19576MD
250mg Each for $76.50

AAL1957603

  SDS
alfa aesar™
L1957603
1g Each for $210.88
Description & Specifications

Specifications

Formula Weight 306.22
Chemical Name or Material cis-Stilbeneboronic acid pinacol ester
MDL Number MFCD03788734
CAS 264144-59-4
Assay 99%
Molecular Formula C20H23BO2
Beilstein 9269561
Synonym cis-(1,2-Diphenylethenyl)boronic acid pinacol cyclic ester
Melting Point 92° to 94°C
Quantity 250mg