Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-Jasmone, 94%

$36.80 - $303.24

Chemical Identifiers

Chemical Name or Material cis-Jasmone
CAS 488-10-8
Molecular Formula C11H16O
Formula Weight 164.25
MDL Number MFCD00001402
Beilstein 1907713
Merck Index 14,5259
Synonym 3-Methyl-2-(2-pentenyl)-2-cyclopenten-1-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1906006

  SDS
alfa aesar™
A1906006
5g Each for $36.80

AAA1906022

  SDS
alfa aesar™
A1906022
100g Each for $303.24
Description & Specifications

Specifications

Formula Weight 164.25
Chemical Name or Material cis-Jasmone
TSCA TSCA
Assay 94%
Molecular Formula C11H16O
MDL Number MFCD00001402
CAS 488-10-8
Beilstein 1907713
Merck Index 14,5259
Synonym 3-Methyl-2-(2-pentenyl)-2-cyclopenten-1-one
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 134° to 135°C (12mmHg)
Quantity 5g
Flash Point 107°C (224°F)
Odor Pleasant
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number GY7301000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.