missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
cis-Epoxysuccinic Acid 97.0+%, TCI America™
Supplier: TCI America E044925G
Specifications
cis-Epoxysuccinic Acid | |
150°C | |
C4H4O5 | |
25 g | |
DCEMCPAKSGRHCN-XIXRPRMCSA-N | |
(2S,3R)-oxirane-2,3-dicarboxylic acid | |
2734802 | |
≥97.0% (GC,T) |
16533-72-5 | |
White | |
MFCD00191645 | |
cis-epoxysuccinic acid, 2r,3s-oxirane-2,3-dicarboxylic acid, cis-oxirane-2,3-dicarboxylic acid, 2,3-oxiranedicarboxylic acid, 2r,3s-rel, 2,3-oxiranedicarboxylic acid, 2r,3s-rel-9ci | |
C1(C(O1)C(=O)O)C(=O)O | |
132.071 | |
132.07 | |
Crystalline Powder |
Chemical Identifiers
16533-72-5 | |
132.071 | |
DCEMCPAKSGRHCN-XIXRPRMCSA-N | |
2734802 | |
C1(C(O1)C(=O)O)C(=O)O |
C4H4O5 | |
MFCD00191645 | |
cis-epoxysuccinic acid, 2r,3s-oxirane-2,3-dicarboxylic acid, cis-oxirane-2,3-dicarboxylic acid, 2,3-oxiranedicarboxylic acid, 2r,3s-rel, 2,3-oxiranedicarboxylic acid, 2r,3s-rel-9ci | |
(2S,3R)-oxirane-2,3-dicarboxylic acid |
Safety and Handling
TSCA : No