Learn More
cis-Dichlorodiamineplatinum(II), 99.99%, (trace metal basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 193760010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisSpecifications
cis-Dichlorodiamineplatinum(II) | |
99.99% | |
White | |
1 g | |
Cl2H6N2Pt | |
MFCD00011623 | |
cis-DDP | |
BSJGASKRWFKGMV-UHFFFAOYSA-L | |
dichloroplatinumbis(ylium) diamine | |
300.05 | |
Trace Metal Basis | |
Glass bottle |
277.0°C | |
15663-27-1 | |
Powder | |
64.0 to 66% (Pt) | |
Pt(NH3)2Cl2 | |
15, 2316 | |
Solubility in water: 0.25g/100mL (25°C) | |
N.N.Cl[Pt++]Cl | |
300.04 | |
99.99% | |
Authentic |
Chemical Identifiers
15663-27-1 | |
300.04 | |
BSJGASKRWFKGMV-UHFFFAOYSA-L | |
dichloroplatinumbis(ylium) diamine |
Cl2H6N2Pt | |
MFCD00011623 | |
cis-DDP | |
N.N.Cl[Pt++]Cl |
Safety and Handling
GHS H Statement
May cause cancer.
Fatal if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If
GHS Signal Word: Danger
EINECSNumber : 239-733-8
RTECSNumber : TP2450000
TSCA : TSCA
RUO – Research Use Only