Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-Dichlorobis(triethylphosphine)platinum(II)

Chemical Identifiers

Chemical Name or Material cis-Dichlorobis(triethylphosphine)platinum(II)
CAS 15692-07-6
Formula Weight 502.32
MDL Number MFCD00058821
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA39284ME

 
alfa aesar™
39284ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 502.32
Chemical Name or Material cis-Dichlorobis(triethylphosphine)platinum(II)
Physical Form Powder
MDL Number MFCD00058821
CAS 15692-07-6
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Melting Point 139° to 142°C
Quantity 500mg
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Hygroscopic
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.