Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-Dichlorobis(pyridine)platinum(II), 99%

$60.66 - $183.18

Chemical Identifiers

Chemical Name or Material cis-Dichlorobis(pyridine)platinum(II)
CAS 14872-21-0
Molecular Formula C10H10Cl2N2Pt
Formula Weight 424.19
MDL Number MFCD00058858
Synonym cis-Bis(pyridine)platinum(II) chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2286501

  SDS
alfa aesar™
B2286501
0.25g Each for $60.66

AAB2286503

  SDS
alfa aesar™
B2286503
1g Each for $183.18
Description & Specifications

Specifications

Formula Weight 424.19
Chemical Name or Material cis-Dichlorobis(pyridine)platinum(II)
Assay 99%
Molecular Formula C10H10Cl2N2Pt
MDL Number MFCD00058858
CAS 14872-21-0
Synonym cis-Bis(pyridine)platinum(II) chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 282° to 288°C
Quantity 0.25g
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.