Learn More
cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 363300010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
cis-Dichlorobis(2, 2'-bipyridine)ruthenium(II) dihydrate | |
Black | |
Authentic | |
Glass bottle | |
MFCD00015504 | |
cis-dichlorobis 2,2'-bipyridine ruthenium ii dihydrate, 99, cis-dichlorobis 2,2'-bipyridine-ruthenium ii dihydrate | |
NHKTUSUPCAKVHT-UHFFFAOYSA-L | |
2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;dihydrate | |
71463877 | |
99% |
15746-57-3 | |
98.5% min. (ex Ru) | |
99% | |
C20H16Cl2N4Ru·2H2O | |
1 g | |
Solubility in water: soluble | |
C1=CC=NC(=C1)C2=CC=CC=N2.C1=CC=NC(=C1)C2=CC=CC=N2.O.O.[Cl-].[Cl-].[Ru+2] | |
520.38 | |
520.38 | |
Crystals or Powder |
Chemical Identifiers
15746-57-3 | |
520.38 | |
NHKTUSUPCAKVHT-UHFFFAOYSA-L | |
71463877 | |
C1=CC=NC(=C1)C2=CC=CC=N2.C1=CC=NC(=C1)C2=CC=CC=N2.O.O.[Cl-].[Cl-].[Ru+2] |
C20H16Cl2N4Ru·2H2O | |
MFCD00015504 | |
cis-dichlorobis 2,2'-bipyridine ruthenium ii dihydrate, 99, cis-dichlorobis 2,2'-bipyridine-ruthenium ii dihydrate | |
2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;dihydrate |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Warning
RUO – Research Use Only