missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-Diamminedichloroplatinum(II), Pt 64.5% min., Alfa Aesar™

Potent Anticancer Agent That Blocks DNA Synthesis.

Manufacturer:  Thermo Fisher Scientific 1047101

 View more versions of this product

Catalog No. AA1047101



Specifications

Specifications

Yellow micro crystals
Odorless
0.25g
15663-27-1
MFCD00011623
300.06
cis-Diamminedichloroplatinum(II)
Yellow
Potent anticancer agent that blocks DNA synthesis
Cl2H6N2Pt
Cisplatin
Microcrystalline

Chemical Identifiers

15663-27-1
Cl2H6N2Pt
MFCD00011623
Cisplatin
Safety and Handling

Safety and Handling

H300-H315-H319-H317-H335-H334-H350: Fatal if swallowed. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer.

DOTInformation : DOT Class: 6.1, Packing Group: II

EINECSNumber : 239-733-8

Documents