Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-Decahydro-1H,6H-3a,5a,8a,10a-tetraazapyrene

$193.64 - $193.64

Chemical Identifiers

Chemical Name or Material cis-Decahydro-1H,6H-3a,5a,8a,10a-tetraazapyrene
CAS 74199-16-9
Molecular Formula C12H22N4
Formula Weight 222.33g/mol
MDL Number MFCD09263305
Synonym cis-Glyoxal-cyclam; Cyclam cis-glyoxal derivative
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2640903

 
alfa aesar™
H26409-03
1g Each for $193.64
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material cis-Decahydro-1H,6H-3a,5a,8a,10a-tetraazapyrene
CAS 74199-16-9
Molecular Formula C12H22N4
Formula Weight 222.33g/mol
MDL Number MFCD09263305
Synonym cis-Glyoxal-cyclam; Cyclam cis-glyoxal derivative
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.