Please login to your online account to display your discounted pricing

cis,cis-μconic acid, 98%, ACROS Organics™


Quantity 2.5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  297760025

 View more versions of this product

Catalog No. AC297760025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 142.11
Physical Form Crystalline Powder
Quantity 2.5g
Color Yellow-beige
Merck Index 15, 6385
Packaging Glass bottle
Chemical Name or Material cis, cis-Muconic acid
Molecular Formula C6H6O4
CAS 1119-72-8
Assay Percent Range 98%
MDL Number MFCD00085266
Infrared Spectrum Authentic
Linear Formula HO2CCH=CHCH=CHCO2H
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material cis, cis-Muconic acid
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H6O4
Linear Formula HO2CCH=CHCH=CHCO2H
Formula Weight 142.11
MDL Number MFCD00085266
Accessories

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents