Please login to your online account to display your discounted pricing

cis,cis,cis,cis-1,2,3,4-Cyclopentanetetracarboxylic Acid 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  111501000

 View more versions of this product

Catalog No. AC111501000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 246.17
Quantity 100g
Color White to yellow-white
EINECS Number 223-256-7
Beilstein 09, IV, 3785
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Water 3% max. (K.F.)
Linear Formula C5H6(CO2H)4
Chemical Name or Material cis,cis,cis,cis-1,2,3,4-Cyclopentanetetracarboxylic acid
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C9H10O8
MDL Number MFCD00001377
CAS 3786-91-2
Appearance of Solution (10% aq. soln.) clear, colorless
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material cis,cis,cis,cis-1,2,3,4-Cyclopentanetetracarboxylic acid
Name Note 98%
CAS 3786-91-2
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H10O8
Linear Formula C5H6(CO2H)4
Formula Weight 246.17
MDL Number MFCD00001377
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents