Please login to your online account to display your discounted pricing

Alfa Aesar™ cis,cis,cis-1,2,3,4-Cyclopentanetetracarboxylic acid, 98%

$26.23 - $47.91

Chemical Identifiers

Chemical Name or Material cis,cis,cis-1,2,3,4-Cyclopentanetetracarboxylic acid
CAS 3786-91-2
Molecular Formula C9H10O8
Formula Weight 246.17
MDL Number MFCD00001377
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2142122

  SDS
alfa aesar™
B2142122
100g Each for $26.23

AAB2142136

  SDS
alfa aesar™
B2142136
500g Each for $47.91
Description & Specifications

Specifications

Formula Weight 246.17
Quantity 100g
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 192° to 195°C (dec.)
CAS 3786-91-2
Chemical Name or Material cis,cis,cis-1,2,3,4-Cyclopentanetetracarboxylic acid
MDL Number MFCD00001377
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C9H10O8
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.