missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis,cis-1,5-Cyclooctadiene, 99%, stab. with 50-200ppm Irganox 1076, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B21196AK

 View more versions of this product

Catalog No. AAB21196AK



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

cis, cis-1, 5-Cyclooctadiene
1552-12-1
0.880
32°C (89°F)
C8H12
MFCD00001752
UN2520
Air Sensitive
VYXHVRARDIDEHS-QGTKBVGQSA-N
(1Z,5Z)-cycloocta-1,5-diene
82916
99%
Stab. With 50-200ppm Irganox 1076
°C to 70°C
151°C to 152°C
0.99
1.494
250 mL
1209288
1,5-cyclooctadiene, cycloocta-1,5-diene, 1z,5z-cycloocta-1,5-diene, 1,5-cyclooctadiene z,z, 1z,5z-cyclooctadiene, z,z-cycloocta-1,5-diene, 1-cis,5-cis-cyclooctadiene, unii-1e1vvd385z, cis,cis-cycloocta-1,5-diene, 1,5-cod
C1CC=CCCC=C1
108.184
108.18
Liquid

Chemical Identifiers

1552-12-1
108.184
VYXHVRARDIDEHS-QGTKBVGQSA-N
82916
C1CC=CCCC=C1
C8H12
MFCD00001752
1,5-cyclooctadiene, cycloocta-1,5-diene, 1z,5z-cycloocta-1,5-diene, 1,5-cyclooctadiene z,z, 1z,5z-cyclooctadiene, z,z-cycloocta-1,5-diene, 1-cis,5-cis-cyclooctadiene, unii-1e1vvd385z, cis,cis-cycloocta-1,5-diene, 1,5-cod
(1Z,5Z)-cycloocta-1,5-diene
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302-H332-H315-H319-H317-H335

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P333+P313-P363-P370+P378q-P501c

H226-H302+H332-H315-H317-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYCLOOCTADIENES

EINECSNumber : 216-291-4

RTECSNumber : GX9620000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only