missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
cis-Bis-(2,2ft.-bipyridine)dichlororuthenium(II) dihydrate, Ru 19% min., Alfa Aesar™
Manufacturer: Alfa Aesar 1277806
Specifications
Black crystalline | |
Black | |
15746-57-3 | |
C20H20Cl2N4O2Ru | |
cis-dichlorobis 2,2'-bipyridine ruthenium ii dihydrate, 99, cis-dichlorobis 2,2'-bipyridine-ruthenium ii dihydrate | |
C1=CC=NC(=C1)C2=CC=CC=N2.C1=CC=NC(=C1)C2=CC=CC=N2.O.O.[Cl-].[Cl-].[Ru+2] | |
520.376 | |
520.38 |
cis-Bis-(2, 2 ft.-bipyridine)dichlororuthenium(II) dihydrate | |
5g | |
Ru 19% min. | |
MFCD00012040 | |
NHKTUSUPCAKVHT-UHFFFAOYSA-L | |
2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;dihydrate | |
71463877 | |
Crystalline |
Chemical Identifiers
15746-57-3 | |
520.376 | |
NHKTUSUPCAKVHT-UHFFFAOYSA-L | |
71463877 | |
C1=CC=NC(=C1)C2=CC=CC=N2.C1=CC=NC(=C1)C2=CC=CC=N2.O.O.[Cl-].[Cl-].[Ru+2] |
C20H20Cl2N4O2Ru | |
MFCD00012040 | |
cis-dichlorobis 2,2'-bipyridine ruthenium ii dihydrate, 99, cis-dichlorobis 2,2'-bipyridine-ruthenium ii dihydrate | |
2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;dihydrate |