Please login to your online account to display your discounted pricing

cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC367500050

 View more versions of this product

Catalog No. AC367500050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥96.0% (ex Ru, on dry substance)
Formula Weight 484.35
Quantity 5g
Color Dark green to black
Water 11% max. (K.F.)
Infrared Spectrum Authentic
Molecular Formula C20H16Cl2N4Ru·xH2O
CAS 98014-14-3
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Glass bottle
Chemical Name or Material cis-Bis(2, 2'-bipyridine)dichlororuthenium(II) hydrate
MDL Number MFCD00150005
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material cis-Bis(2, 2'-bipyridine)dichlororuthenium(II) hydrate
CAS 98014-14-3
Assay ≥96.0% (ex Ru, on dry substance)
Assay Percent Range 97%
Molecular Formula C20H16Cl2N4Ru·xH2O
Formula Weight 484.35
MDL Number MFCD00150005
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents