Please login to your online account to display your discounted pricing

cis-ACPD, Tocris


Potent NMDA agonist; Also group II mGluR agonist

Quantity 10mg

Manufacturer: r & d systems  RDS018610

 View more versions of this product

Catalog No. 018610

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 173.17
CAS 477331-06-9
Assay Percent Range >98%
Molecular Formula C7H11NO4
Recommended Storage Store at Room Temperature
Synonym (±)-1-Aminocyclopentane-cis-1, 3-dicarboxylic acid
Chemical Name or Material cis-ACPD

Specifications

Chemical Name or Material cis-ACPD
Assay Percent Range >98%
Molecular Formula C7H11NO4
Formula Weight 173.17
Synonym (±)-1-Aminocyclopentane-cis-1, 3-dicarboxylic acid
Accessories

Store at Room Temperature