Please login to your online account to display your discounted pricing

cis-ACPD, Tocris

Potent NMDA agonist; Also group II mGluR agonist

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material cis-ACPD
Assay Percent Range >98%
Molecular Formula C7H11NO4
Formula Weight 173.17
Synonym (±)-1-Aminocyclopentane-cis-1, 3-dicarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

018650

 
r & d systems
RDS018650
50mg Each for $904.70

018610

 
r & d systems
RDS018610
10mg Each for $221.01
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 173.17
Chemical Name or Material cis-ACPD
Assay Percent Range >98%
Molecular Formula C7H11NO4
CAS 477331-06-9
Synonym (±)-1-Aminocyclopentane-cis-1, 3-dicarboxylic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature