Please login to your online account to display your discounted pricing

cis-ACBD, Tocris


Potent, selective L-glutamate uptake inhibitor

Quantity 50mg

Manufacturer: r & d systems  RDS027150

 View more versions of this product

Catalog No. 027150

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material cis-ACBD
CAS 73550-55-7
Recommended Storage Store at Room Temperature
Quantity 50mg
Assay Percent Range >98%
Molecular Formula C6H9NO4
Formula Weight 159.14
Synonym cis-1-Aminocyclobutane-1, 3-dicarboxylic acid

Specifications

Chemical Name or Material cis-ACBD
Assay Percent Range >98%
Molecular Formula C6H9NO4
Formula Weight 159.14
Synonym cis-1-Aminocyclobutane-1, 3-dicarboxylic acid
Accessories

Store at Room Temperature