Please login to your online account to display your discounted pricing

cis-ACBD, Tocris

Potent, selective L-glutamate uptake inhibitor

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material cis-ACBD
Assay Percent Range >98%
Molecular Formula C6H9NO4
Formula Weight 159.14
Synonym cis-1-Aminocyclobutane-1, 3-dicarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

027110

 
r & d systems
RDS027110
10mg Each for $177.37

027150

 
r & d systems
RDS027150
50mg Each for $746.48
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 159.14
CAS 73550-55-7
Synonym cis-1-Aminocyclobutane-1, 3-dicarboxylic acid
Chemical Name or Material cis-ACBD
Assay Percent Range >98%
Molecular Formula C6H9NO4
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature