Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-9-Tricosene, 96%

$269.38 - $269.38

Chemical Identifiers

Chemical Name or Material cis-9-Tricosene
CAS 27519-02-4
Molecular Formula C23H46
Formula Weight 322.62
MDL Number MFCD00008988
Beilstein 1841622
Merck Index 14,6309
Synonym Muscalure
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0961506

 
alfa aesar™
L0961506
5g Each for $269.38
Description & Specifications

Specifications

Formula Weight 322.62
Chemical Name or Material cis-9-Tricosene
Assay 96%
Molecular Formula C23H46
MDL Number MFCD00008988
CAS 27519-02-4
TSCA TSCA
Beilstein 1841622
Merck Index 14,6309
Synonym Muscalure
Boiling Point 299° to 300°C
Quantity 5g
Flash Point >110°C (230°F)
RTECS Number YD0807000

Applications
cis-9-Tricosene acts as an insect attractant (sex pheromone) of the female house fly. Further, it is used for biochemical research. In addition, it is used in fly paper and other traps to lure male flies.

Solubility
Miscible with water and hexane. Immiscible with alcohol.

Notes
Incompatible with strong oxidizing agents.