missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-9-Tricosene, 96%

Manufacturer:  Alfa Aesar™ L0961506

 View more versions of this product

Catalog No. AAL0961506



Description

Description

Applications
cis-9-Tricosene acts as an insect attractant (sex pheromone) of the female house fly. Further, it is used for biochemical research. In addition, it is used in fly paper and other traps to lure male flies.

Solubility
Miscible with water and hexane. Immiscible with alcohol.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

cis-9-Tricosene
299°C to 300°C
27519-02-4
MFCD00008988
14,6309
IGOWHGRNPLFNDJ-ZPHPHTNESA-N
(Z)-tricos-9-ene
5365075
96%
>110°C (230°F)
5g
C23H46
1841622
cis-9-tricosene, muscalure, z-9-tricosene, z-tricos-9-ene, 9-tricosene, z, muscamone, cis-tricos-9-ene, caswell no. 883c, 9z-tricosene, unii-6bsp6hfw73
CCCCCCCCCCCCCC=CCCCCCCCC
322.621
322.62

Chemical Identifiers

27519-02-4
322.621
IGOWHGRNPLFNDJ-ZPHPHTNESA-N
5365075
CCCCCCCCCCCCCC=CCCCCCCCC
C23H46
MFCD00008988
cis-9-tricosene, muscalure, z-9-tricosene, z-tricos-9-ene, 9-tricosene, z, muscamone, cis-tricos-9-ene, caswell no. 883c, 9z-tricosene, unii-6bsp6hfw73
(Z)-tricos-9-ene
Safety and Handling

Safety and Handling

RTECSNumber : YD0807000

TSCA : TSCA

Documents