Please login to your online account to display your discounted pricing

cis-8-Azabicyclo[4.3.0]nonane, 97%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC444000050

 View more versions of this product

Catalog No. AC444000050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥96.0%
Formula Weight 125.22
Quantity 5g
Color Clear pale brown to red-brown
Chemical Name or Material cis-8-Azabicyclo[4.3.0]nonane
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C8H15N
CAS 1470-99-1
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Danger

Specifications

Chemical Name or Material cis-8-Azabicyclo[4.3.0]nonane
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H15N
Formula Weight 125.22
Accessories

Danger

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.