missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-8-Azabicyclo[4.3.0]nonane, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444000010

 View more versions of this product

Catalog No. AC444000010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
125.22
Clear pale brown to red-brown
1470-99-1
97%
Liquid
1g
C8H15N
cis-8-Azabicyclo[4.3.0]nonane
Authentic

Chemical Identifiers

97%
125.22
≥96.0%
C8H15N
cis-8-Azabicyclo[4.3.0]nonane

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger