Please login to your online account to display your discounted pricing

cis-8-Azabicyclo[4.3.0]nonane, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material cis-8-Azabicyclo[4.3.0]nonane
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H15N
Formula Weight 125.22
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC444000010

 
thermo fisher scientific
AC444000010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC444000050

 
thermo fisher scientific
AC444000050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 125.22
Quantity 1g
Color Clear pale brown to red-brown
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C8H15N
CAS 1470-99-1
Chemical Name or Material cis-8-Azabicyclo[4.3.0]nonane
Assay Percent Range 97%
Infrared Spectrum Authentic
Safety and Handling

Danger

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.