Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-6-(Benzyloxycarbonylamino)-3-cyclohexene-1-carboxylic acid, 97%

Chemical Identifiers

Chemical Name or Material cis-6-(Benzyloxycarbonylamino)-3-cyclohexene-1-carboxylic acid
CAS 124753-65-7
Formula Weight 275.29
MDL Number MFCD01863246
Synonym cis-6-Benzyloxycarbonylaminocyclohex-3-enecarboxylic acid; Z-1,2-cis-ACHeC-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6265003

 
alfa aesar™
H6265003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62650MD

 
alfa aesar™
H62650MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 275.29
Quantity 1g
Chemical Name or Material cis-6-(Benzyloxycarbonylamino)-3-cyclohexene-1-carboxylic acid
Physical Form Solid
Assay 97%
MDL Number MFCD01863246
CAS 124753-65-7
Synonym cis-6-Benzyloxycarbonylaminocyclohex-3-enecarboxylic acid; Z-1,2-cis-ACHeC-OH