Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-5-Octen-1-ol, 95%, remainder mainly trans-isomer

$68.50 - $246.28

Chemical Identifiers

Chemical Name or Material cis-5-Octen-1-ol
CAS 64275-73-6
Molecular Formula C8H16O
Formula Weight 128.22
MDL Number MFCD00015569
Beilstein 1840670
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1945414

  SDS
alfa aesar™
A1945414
25g Each for $246.28

AAA1945406

  SDS
alfa aesar™
A1945406
5g Each for $68.50
Description & Specifications

Specifications

Formula Weight 128.22
Chemical Name or Material cis-5-Octen-1-ol
TSCA TSCA
Assay 95%
Assay Percent Notes remainder mainly trans-isomer
Molecular Formula C8H16O
MDL Number MFCD00015569
CAS 64275-73-6
Beilstein 1840670
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Boiling Point 95°C (25mmHg)
Quantity 25g
Flash Point 82°C (179°F)
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.