missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-5-Norbornene-exo-2,3-dicarboxylic anhydride, 95%

Manufacturer:  Alfa Aesar™ H5344614

 View more versions of this product

Catalog No. AAH5344614



Description

Description

Applications
cis-5-Norbornene-exo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

cis-5-Norbornene-exo-2,3-dicarboxylic anhydride
25g
C9H8O3
1r,2r,6s,7s-4-oxatricyclo 5.2.1.0 2 ,? dec-8-ene-3,5-dione
C1C2C=CC1C3C2C(=O)OC3=O
637794
95%
140°C to 145°C
2746-19-2
MFCD00630580
KNDQHSIWLOJIGP-RNGGSSJXSA-N
164.16
164.16

Chemical Identifiers

2746-19-2
164.16
KNDQHSIWLOJIGP-RNGGSSJXSA-N
637794
C9H8O3
MFCD00630580
1r,2r,6s,7s-4-oxatricyclo 5.2.1.0 2 ,? dec-8-ene-3,5-dione
C1C2C=CC1C3C2C(=O)OC3=O
Safety and Handling

Safety and Handling

GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.

P261-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P310-P333+P313-P342+P311-P363-P501c

H317-H318-H334-H335

EINECSNumber : 220-384-5

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents